missing translation for 'onlineSavingsMsg'
Learn More

1-BOC-3-aminoazetidine, 95%

Catalog No. AC451180050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC451180050 5 g
AC451180010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC451180050 Supplier Thermo Scientific Chemicals Supplier No. 451180050
Only null left

CAS: 193269-78-2 | C8H16N2O2 | 172.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 193269-78-2
Molecular Formula C8H16N2O2
Molecular Weight (g/mol) 172.23
InChI Key WPGLRFGDZJSQGI-UHFFFAOYSA-N
Synonym 1-boc-3-amino azetidine, 1-boc-3-aminoazetidine, 3-amino-1-n-boc-azetidine, 1-n-boc-3-aminoazetidine, 3-amino-1-boc-azetidine, 3-amino-1-n-boc azetidine, tert-butyl 3-aminoazetidinecarboxylate, 1-azetidinecarboxylic acid, 3-amino-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-aminoazetidine, 3-amino-1-tert-butoxycarbonyl azetidine
PubChem CID 1516507
IUPAC Name tert-butyl 3-aminoazetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(C1)N

Specifications

Assay Percent Range 94% min. (GC)
Quantity 5 g
Formula Weight 172.23
Percent Purity 95%
Chemical Name or Material 1-BOC-3-aminoazetidine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

RUO – Research Use Only