missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-3-aminoazetidine, 94%

CAS: 193269-78-2 | C8H16N2O2 | 172.228 g/mol

Supplier:  Thermo Scientific Chemicals H2821803

 View more versions of this product

Catalog No. AAH2821803


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Boc-3-aminoazetidine
C8H16N2O2
MFCD01861753
1-boc-3-amino azetidine, 1-boc-3-aminoazetidine, 3-amino-1-n-boc-azetidine, 1-n-boc-3-aminoazetidine, 3-amino-1-boc-azetidine, 3-amino-1-n-boc azetidine, tert-butyl 3-aminoazetidinecarboxylate, 1-azetidinecarboxylic acid, 3-amino-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-aminoazetidine, 3-amino-1-tert-butoxycarbonyl azetidine
CC(C)(C)OC(=O)N1CC(C1)N
1 g
1516507
Air Sensitive
193269-78-2
1.465
UN2735
WPGLRFGDZJSQGI-UHFFFAOYSA-N
tert-butyl 3-aminoazetidine-1-carboxylate
172.228
172.23
94%

Chemical Identifiers

193269-78-2
172.228
WPGLRFGDZJSQGI-UHFFFAOYSA-N
1516507
CC(C)(C)OC(=O)N1CC(C1)N
C8H16N2O2
MFCD01861753
1-boc-3-amino azetidine, 1-boc-3-aminoazetidine, 3-amino-1-n-boc-azetidine, 1-n-boc-3-aminoazetidine, 3-amino-1-boc-azetidine, 3-amino-1-n-boc azetidine, tert-butyl 3-aminoazetidinecarboxylate, 1-azetidinecarboxylic acid, 3-amino-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-3-aminoazetidine, 3-amino-1-tert-butoxycarbonyl azetidine
tert-butyl 3-aminoazetidine-1-carboxylate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only