missing translation for 'onlineSavingsMsg'
Learn More

1-Benzylpiperidine 98.0+%, TCI America™

Catalog No. p-7123694
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
5 mL
Catalog No. Quantity
B46985ML 5 mL
B469825ML 25 mL
2 options

Catalog No. B46985ML

Supplier: TCI America B46985ML

Only null left

Chemical Identifiers

CAS 2905-56-8
Molecular Formula C12H18ClN
Molecular Weight (g/mol) 211.73
MDL Number MFCD00224901
InChI Key MPPIBJJDFLONMO-UHFFFAOYSA-N
Synonym piperidine, 1-phenylmethyl, benzylpiperidine, unii-7hze16210b, n-benzylpiperidine, benzyl-piperidine, n-benzyl piperidine, n-benzyl-piperidine, 1-benzyl-piperidine, acmc-20a2pl
PubChem CID 76190
IUPAC Name hydrogen 1-benzylpiperidine chloride
SMILES [H+].[Cl-].C(N1CCCCC1)C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 66°C
Quantity 5 mL
Formula Weight 175.28
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Benzylpiperidine