missing translation for 'onlineSavingsMsg'
Learn More

1-Benzylpiperidine, 98%

Catalog No. p-7047501
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1933909 10 g
AAL1933918 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1933909 Supplier Thermo Scientific Chemicals Supplier No. L1933909

CAS: 2905-56-8 | C12H18ClN | 211.73 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2905-56-8
Molecular Formula C12H18ClN
Molecular Weight (g/mol) 211.73
MDL Number MFCD00224901
InChI Key MPPIBJJDFLONMO-UHFFFAOYSA-N
Synonym piperidine, 1-phenylmethyl, benzylpiperidine, unii-7hze16210b, n-benzylpiperidine, benzyl-piperidine, n-benzyl piperidine, n-benzyl-piperidine, 1-benzyl-piperidine, acmc-20a2pl
PubChem CID 76190
SMILES [H+].[Cl-].C(N1CCCCC1)C1=CC=CC=C1

Specifications

Density 0.95
Boiling Point 120°C to 123°C (9 mmHg)
Refractive Index 1.526
Quantity 10 g
Beilstein 5639
IUPAC Name 1-benzylpiperidine
Formula Weight 175.28
Percent Purity 98%
Chemical Name or Material 1-Benzylpiperidine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 220-809-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only