Learn More
1-Benzyloxycarbonylpyrrolidine-3-carboxaldehyde, 97%
CAS: 276872-86-7 | C13H15NO3 | 233.267 g/mol
$178.30 - $593.14
Chemical Identifiers
| CAS | 276872-86-7 |
|---|---|
| Molecular Formula | C13H15NO3 |
| Molecular Weight (g/mol) | 233.267 |
| MDL Number | MFCD05861538 |
| InChI Key | GDPSCBPOCONUDM-UHFFFAOYSA-N |
| Synonym | 1-n-cbz-3-formyl-pyrrolidine, 1-cbz-3-pyrrolidinecarbaldehyde, 3-formyl-pyrrolidine-1-carboxylic acid benzyl ester, 1-cbz-pyrrolidine-3-carboxaldehyde, 1-benzyloxycarbonylpyrrolidine-3-carboxaldehyde, 3-formyl-pyrrolidine-1-carboxylicacidbenzylester, benzyl 3-formyl-1-pyrrolidinecarboxylate, 1-pyrrolidinecarboxylic acid, 3-formyl-, phenylmethyl ester, z-3-formylpyrrolidine, n-cbz-3-formyl pyrrolidine |
| PubChem CID | 4620242 |
| IUPAC Name | benzyl 3-formylpyrrolidine-1-carboxylate |
| SMILES | C1CN(CC1C=O)C(=O)OCC2=CC=CC=C2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2811703
|
Thermo Scientific Chemicals
H2811703 |
1 g |
Each for $178.30
|
|
|||||
|
AAH2811706
|
Thermo Scientific Chemicals
H2811706 |
5 g |
Each for $593.14
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 276872-86-7 | |
| 233.267 | |
| GDPSCBPOCONUDM-UHFFFAOYSA-N | |
| 4620242 | |
| C1CN(CC1C=O)C(=O)OCC2=CC=CC=C2 |
| C13H15NO3 | |
| MFCD05861538 | |
| 1-n-cbz-3-formyl-pyrrolidine, 1-cbz-3-pyrrolidinecarbaldehyde, 3-formyl-pyrrolidine-1-carboxylic acid benzyl ester, 1-cbz-pyrrolidine-3-carboxaldehyde, 1-benzyloxycarbonylpyrrolidine-3-carboxaldehyde, 3-formyl-pyrrolidine-1-carboxylicacidbenzylester, benzyl 3-formyl-1-pyrrolidinecarboxylate, 1-pyrrolidinecarboxylic acid, 3-formyl-, phenylmethyl ester, z-3-formylpyrrolidine, n-cbz-3-formyl pyrrolidine | |
| benzyl 3-formylpyrrolidine-1-carboxylate |
Specifications
| 276872-86-7 | |
| MFCD05861538 | |
| Air Sensitive | |
| GDPSCBPOCONUDM-UHFFFAOYSA-N | |
| benzyl 3-formylpyrrolidine-1-carboxylate | |
| 4620242 | |
| 97% |
| C13H15NO3 | |
| 1 g | |
| 1-n-cbz-3-formyl-pyrrolidine, 1-cbz-3-pyrrolidinecarbaldehyde, 3-formyl-pyrrolidine-1-carboxylic acid benzyl ester, 1-cbz-pyrrolidine-3-carboxaldehyde, 1-benzyloxycarbonylpyrrolidine-3-carboxaldehyde, 3-formyl-pyrrolidine-1-carboxylicacidbenzylester, benzyl 3-formyl-1-pyrrolidinecarboxylate, 1-pyrrolidinecarboxylic acid, 3-formyl-, phenylmethyl ester, z-3-formylpyrrolidine, n-cbz-3-formyl pyrrolidine | |
| C1CN(CC1C=O)C(=O)OCC2=CC=CC=C2 | |
| 233.267 | |
| 233.27 | |
| 1-Benzyloxycarbonylpyrrolidine-3-carboxaldehyde |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only