missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyloxycarbonylpyrrolidine-3-carboxaldehyde, 97%

Catalog No. AAH2811703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2811703 1 g
AAH2811706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2811703 Supplier Thermo Scientific Chemicals Supplier No. H2811703

CAS: 276872-86-7 | C13H15NO3 | 233.267 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 276872-86-7
Molecular Formula C13H15NO3
Molecular Weight (g/mol) 233.267
MDL Number MFCD05861538
InChI Key GDPSCBPOCONUDM-UHFFFAOYSA-N
Synonym 1-n-cbz-3-formyl-pyrrolidine, 1-cbz-3-pyrrolidinecarbaldehyde, 3-formyl-pyrrolidine-1-carboxylic acid benzyl ester, 1-cbz-pyrrolidine-3-carboxaldehyde, 1-benzyloxycarbonylpyrrolidine-3-carboxaldehyde, 3-formyl-pyrrolidine-1-carboxylicacidbenzylester, benzyl 3-formyl-1-pyrrolidinecarboxylate, 1-pyrrolidinecarboxylic acid, 3-formyl-, phenylmethyl ester, z-3-formylpyrrolidine, n-cbz-3-formyl pyrrolidine
PubChem CID 4620242
IUPAC Name benzyl 3-formylpyrrolidine-1-carboxylate
SMILES C1CN(CC1C=O)C(=O)OCC2=CC=CC=C2

Specifications

Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 233.27
Percent Purity 97%
Chemical Name or Material 1-Benzyloxycarbonylpyrrolidine-3-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only