Learn More
1-Benzyloxycarbonyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals
$312.47 - $312.47
Chemical Identifiers
| CAS | 286961-15-7 |
|---|---|
| Molecular Formula | C19H26BNO4 |
| Molecular Weight (g/mol) | 343.23 |
| MDL Number | MFCD11521562 |
| InChI Key | QDSFHRPYZPQWEJ-UHFFFAOYSA-N |
| Synonym | benzyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-5,6-dihydropyridine-1 2h-carboxylate, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylic acid benzyl ester, n-cbz-3,6-dihydro-2h-pyridine-4-boronic acid pinacol ester, benzyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylate, 1-benzyloxycarbonyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, benzyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydropyridine-1 2h-carboxylate, benzyl 5,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-1 2h-carboxylate, n-cbz-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, benzyl 4-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylate, benzyl 4-4,4,5,5-tetrameth |
| PubChem CID | 11290836 |
| IUPAC Name | benzyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyridine-1-carboxylate |
| SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CCN(CC2)C(=O)OCC3=CC=CC=C3 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6489103
|
Thermo Scientific Chemicals
H6489103 |
1 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 286961-15-7 | |
| 343.23 | |
| QDSFHRPYZPQWEJ-UHFFFAOYSA-N | |
| 11290836 | |
| B1(OC(C(O1)(C)C)(C)C)C2=CCN(CC2)C(=O)OCC3=CC=CC=C3 |
| C19H26BNO4 | |
| MFCD11521562 | |
| benzyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-5,6-dihydropyridine-1 2h-carboxylate, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylic acid benzyl ester, n-cbz-3,6-dihydro-2h-pyridine-4-boronic acid pinacol ester, benzyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylate, 1-benzyloxycarbonyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, benzyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydropyridine-1 2h-carboxylate, benzyl 5,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-1 2h-carboxylate, n-cbz-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, benzyl 4-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylate, benzyl 4-4,4,5,5-tetrameth | |
| benzyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyridine-1-carboxylate |
Specifications
| 286961-15-7 | |
| White | |
| MFCD11521562 | |
| 8571588 | |
| QDSFHRPYZPQWEJ-UHFFFAOYSA-N | |
| benzyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyridine-1-carboxylate | |
| 11290836 | |
| 98% | |
| 1-Benzyloxycarbonyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester |
| 88°C to 90°C | |
| C19H26BNO4 | |
| 1 g | |
| benzyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-5,6-dihydropyridine-1 2h-carboxylate, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylic acid benzyl ester, n-cbz-3,6-dihydro-2h-pyridine-4-boronic acid pinacol ester, benzyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylate, 1-benzyloxycarbonyl-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, benzyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydropyridine-1 2h-carboxylate, benzyl 5,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine-1 2h-carboxylate, n-cbz-1,2,3,6-tetrahydropyridine-4-boronic acid pinacol ester, benzyl 4-tetramethyl-1,3,2-dioxaborolan-2-yl-3,6-dihydro-2h-pyridine-1-carboxylate, benzyl 4-4,4,5,5-tetrameth | |
| B1(OC(C(O1)(C)C)(C)C)C2=CCN(CC2)C(=O)OCC3=CC=CC=C3 | |
| 343.23 | |
| 343.23 | |
| Solid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only