missing translation for 'onlineSavingsMsg'
Learn More

1-Benzylindole, 97%

Catalog No. AAH6341403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6341403 1 g
AAH6341406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6341403 Supplier Thermo Scientific Chemicals Supplier No. H6341403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3377-71-7 | C15H13N | 207.276 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3377-71-7
Molecular Formula C15H13N
Molecular Weight (g/mol) 207.276
MDL Number MFCD00015884
InChI Key NJZQOCCEDXRQJM-UHFFFAOYSA-N
Synonym 1-benzyl-1h-indole, n-benzylindole, 1-benzyl-indole, 1h-indole, 1-phenylmethyl, n-benzyl indole, n-benzyl-indole, zlchem 819, phenylmethyl-1h-indole, cambridge id 5106611, 1h-indole, phenylmethyl
PubChem CID 96913
IUPAC Name 1-benzylindole
SMILES C1=CC=C(C=C1)CN2C=CC3=CC=CC=C32

Specifications

Color White to Yellow
Quantity 1 g
Beilstein 148884
Formula Weight 207.27
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Benzylindole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only