missing translation for 'onlineSavingsMsg'
Learn More

1-Benzylindole-3-carboxylic acid, 95%

Catalog No. p-7057016
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6629403 1 g
AAH6629406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6629403 Supplier Thermo Scientific Chemicals Supplier No. H6629403

CAS: 27018-76-4 | C16H13NO2 | 251.285 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27018-76-4
Molecular Formula C16H13NO2
Molecular Weight (g/mol) 251.285
MDL Number MFCD00057094
InChI Key LVYDDRHDOKXFMW-UHFFFAOYSA-N
Synonym 1-benzyl-1h-indole-3-carboxylic acid, indole-3-carboxylic acid, 1-benzyl, 1h-indole-3-carboxylic acid, 1-phenylmethyl, pubchem7313, cbmicro_033327, cambridge id 5791025, 5-22-03-00037 beilstein handbook reference, 1-benzylindol-3-carboxylic acid, n-benzylindole-3-carboxylic acid, 1h-indole-3-carboxylicacid, 1-phenylmethyl
PubChem CID 33671
IUPAC Name 1-benzylindole-3-carboxylic acid
SMILES C1=CC=C(C=C1)CN2C=C(C3=CC=CC=C32)C(=O)O

Specifications

Quantity 1 g
Beilstein 404957
Formula Weight 251.29
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 1-Benzylindole-3-carboxylic acid
RTECSNumber NL5995800
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only