missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Benzyl N-(tert-Butoxycarbonyl)-L-aspartate 98.0+%, TCI America™
$68.97 - $270.29
Chemical Identifiers
| CAS | 30925-18-9 |
|---|---|
| Molecular Formula | C16H21NO6 |
| Molecular Weight (g/mol) | 323.345 |
| InChI Key | LDRWTKQWSXGSTM-LBPRGKRZSA-N |
| Synonym | boc-asp-obzl, boc-l-aspartic acid 1-benzyl ester, 1-benzyl n-boc-l-aspartate, n-tert-butoxycarbonyl-l-aspartic acid 1-benzyl ester, 3s-4-benzyloxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, 1-benzyl n-tert-butoxycarbonyl-l-aspartate, commericial, 3s-3-tert-butoxy carbonylamino-3-benzyloxycarbonyl propanoic acid, pubchem12927, boc-l-asp h-obn |
| PubChem CID | 7017887 |
| IUPAC Name | (3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid |
| SMILES | CC(C)(C)OC(=O)NC(CC(=O)O)C(=O)OCC1=CC=CC=C1 |
Chemical Identifiers
| 30925-18-9 | |
| 323.345 | |
| boc-asp-obzl, boc-l-aspartic acid 1-benzyl ester, 1-benzyl n-boc-l-aspartate, n-tert-butoxycarbonyl-l-aspartic acid 1-benzyl ester, 3s-4-benzyloxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, 1-benzyl n-tert-butoxycarbonyl-l-aspartate, commericial, 3s-3-tert-butoxy carbonylamino-3-benzyloxycarbonyl propanoic acid, pubchem12927, boc-l-asp h-obn | |
| (3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid |
| C16H21NO6 | |
| LDRWTKQWSXGSTM-LBPRGKRZSA-N | |
| 7017887 | |
| CC(C)(C)OC(=O)NC(CC(=O)O)C(=O)OCC1=CC=CC=C1 |
Specifications
| 30925-18-9 | |
| White | |
| 5 g | |
| LDRWTKQWSXGSTM-LBPRGKRZSA-N | |
| (3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid | |
| 7017887 | |
| ≥98.0% (HPLC,T) | |
| 1-Benzyl N-(tert-Butoxycarbonyl)-L-aspartate |
| 99°C | |
| C16H21NO6 | |
| boc-asp-obzl, boc-l-aspartic acid 1-benzyl ester, 1-benzyl n-boc-l-aspartate, n-tert-butoxycarbonyl-l-aspartic acid 1-benzyl ester, 3s-4-benzyloxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid, 1-benzyl n-tert-butoxycarbonyl-l-aspartate, commericial, 3s-3-tert-butoxy carbonylamino-3-benzyloxycarbonyl propanoic acid, pubchem12927, boc-l-asp h-obn | |
| CC(C)(C)OC(=O)NC(CC(=O)O)C(=O)OCC1=CC=CC=C1 | |
| 323.345 | |
| 323.35 | |
| Crystalline Powder |
Safety and Handling
TSCA : No