missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl N-Carbobenzoxy-L-glutamate 98.0+%, TCI America™

Catalog No. B39895G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39895G 5 g
B398925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39895G Supplier TCI America Supplier No. B39895G

Chemical Identifiers

CAS 3705-42-8
Molecular Formula C20H21NO6
Molecular Weight (g/mol) 371.389
MDL Number MFCD00077013
InChI Key VWHKODOUMSMUAF-KRWDZBQOSA-N
Synonym z-glu-obzl, cbz-glu-obzl, cbz-l-glutamic acid 1-benzyl ester, z-l-glu-obzl, n-cbz-l-glutamic acid benzyl ester, n-carbobenzyloxy-l-glutamic acid o-benzyl ester, s-5-benzyloxy-4-benzyloxy carbonyl amino-5-oxopentanoic acid, 4s-5-benzyloxy-4-benzyloxy carbonyl amino-5-oxopentanoic acid, n-benzyloxycarbonyl-l-glutamic acid, a-benzyl ester, z-l-glutamic acid alpha-benzyl ester
PubChem CID 6994031
IUPAC Name (4S)-5-oxo-5-phenylmethoxy-4-(phenylmethoxycarbonylamino)pentanoic acid
SMILES C1=CC=C(C=C1)COC(=O)C(CCC(=O)O)NC(=O)OCC2=CC=CC=C2

Specifications

Melting Point 100°C
Color White
Quantity 5 g
Formula Weight 371.39
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzyl N-Carbobenzoxy-L-glutamate