missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl N-Carbobenzoxy-D-glutamate 98.0+%, TCI America™

Catalog No. B42815G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B42815G Supplier TCI America Supplier No. B42815G

Chemical Identifiers

CAS 65706-99-2
Molecular Formula C20H21NO6
Molecular Weight (g/mol) 371.389
MDL Number MFCD00069647
InChI Key VWHKODOUMSMUAF-QGZVFWFLSA-N
Synonym z-d-glu-obzl, cbz-d-glu-obzl, n-cbz-d-glutamic acid alpha-benzyl ester, r-5-benzyloxy-4-benzyloxy carbonyl amino-5-oxopentanoic acid, 1-benzyl n-carbobenzoxy-d-glutamate, 4r-5-benzyloxy-4-benzyloxy carbonyl amino-5-oxopentanoic acid, z-d-glu-obn, cbz-d-glu-och2ph, pubchem14966, cbz-d-glu oh-obzl
PubChem CID 7018162
IUPAC Name (4R)-5-oxo-5-phenylmethoxy-4-(phenylmethoxycarbonylamino)pentanoic acid
SMILES C1=CC=C(C=C1)COC(=O)C(CCC(=O)O)NC(=O)OCC2=CC=CC=C2

Specifications

Melting Point 100°C
Color White
Quantity 5 g
Formula Weight 371.39
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzyl N-Carbobenzoxy-D-glutamate