missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl L-Glutamate 98.0+%, TCI America™

Catalog No. B29965G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29965G 5 g
B29961G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29965G Supplier TCI America Supplier No. B29965G

Chemical Identifiers

CAS 13030-09-6
Molecular Formula C12H15NO4
Molecular Weight (g/mol) 237.255
MDL Number MFCD00037312
InChI Key HFZKKJHBHCZXTQ-JTQLQIEISA-N
Synonym L-Glutamic Acid 1-Benzyl Ester, H-Glu-OBzl
PubChem CID 7009632
IUPAC Name (4S)-4-amino-5-oxo-5-phenylmethoxypentanoic acid
SMILES C1=CC=C(C=C1)COC(=O)C(CCC(=O)O)N

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 237.26
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzyl L-Glutamate