missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl D-Glutamate 98.0+%, TCI America™

Catalog No. B29955G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29955G 5 g
B29951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29955G Supplier TCI America Supplier No. B29955G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 79338-14-0
Molecular Formula C12H15NO4
Molecular Weight (g/mol) 237.255
MDL Number MFCD00237065
InChI Key HFZKKJHBHCZXTQ-SNVBAGLBSA-N
Synonym h-d-glu-obzl, 1-benzyl d-glutamate, r-4-amino-5-benzyloxy-5-oxopentanoic acid, d-glutamic acid 1-benzyl ester, 7-benzyl glutamate, 4r-4-amino-5-benzyloxy-5-oxopentanoic acid
PubChem CID 7019728
IUPAC Name (4R)-4-amino-5-oxo-5-phenylmethoxypentanoic acid
SMILES C1=CC=C(C=C1)COC(=O)C(CCC(=O)O)N

Specifications

Melting Point 144°C
Color White
Quantity 5 g
Formula Weight 237.26
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzyl D-Glutamate