missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-5-bromoindole, 97%

CAS: 10075-51-1 | C15H12BrN | 286.17 g/mol

$125.86 - $483.70

Chemical Identifiers

CAS 10075-51-1
Molecular Formula C15H12BrN
Molecular Weight (g/mol) 286.17
MDL Number MFCD04337704
InChI Key AQXJFUYUNHLBGU-UHFFFAOYSA-N
Synonym 1-benzyl-5-bromo-1h-indole, n-benzyl-5-bromoindole
PubChem CID 285756
IUPAC Name 1-benzyl-5-bromo-1H-indole
SMILES BrC1=CC=C2N(CC3=CC=CC=C3)C=CC2=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5767603
View Documents
Thermo Scientific Chemicals
H5767603
1 g
Each for $125.86
Only null left
 
AAH5767606
View Documents
Thermo Scientific Chemicals
H5767606
5 g
Each for $483.70
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10075-51-1
286.17
AQXJFUYUNHLBGU-UHFFFAOYSA-N
285756
BrC1=CC=C2N(CC3=CC=CC=C3)C=CC2=C1
C15H12BrN
MFCD04337704
1-benzyl-5-bromo-1h-indole, n-benzyl-5-bromoindole
1-benzyl-5-bromo-1H-indole

Specifications

10075-51-1
MFCD04337704
1-benzyl-5-bromo-1h-indole, n-benzyl-5-bromoindole
BrC1=CC=C2N(CC3=CC=CC=C3)C=CC2=C1
286.17
286.17
1-Benzyl-5-bromoindole
C15H12BrN
1 g
AQXJFUYUNHLBGU-UHFFFAOYSA-N
1-benzyl-5-bromo-1H-indole
285756
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only