missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-4-cyano-4-phenylpiperidine Hydrochloride 98.0+%, TCI America™

Catalog No. b43935g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B43935G 5 g
B439325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B43935G Supplier TCI America Supplier No. B43935G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 71258-18-9
Molecular Formula C19H21ClN2
Molecular Weight (g/mol) 312.841
MDL Number MFCD00035361
InChI Key SPXFNRMZPRHYRH-UHFFFAOYSA-N
Synonym 1-benzyl-4-cyano-4-phenylpiperidine hydrochloride, 1-benzyl-4-phenylpiperidine-4-carbonitrile hydrochloride, 1-benzyl-4-cyano-4-phenylpiperidine hcl, 1-n-benzyl-4-cyano-4-phenylpiperidine hydrochloride, n-benzyl-4-cyano-4-phenylpiperidine hydrochloride, 4-phenyl-1-benzylpiperidine-4-carbonitrile, chloride, 4-phenyl-1-phenylmethyl-4-piperidinecarbonitrile hydrochloride, pubchem13098, acmc-1bfv8, c19h20n2.clh
PubChem CID 12652062
IUPAC Name 1-benzyl-4-phenylpiperidine-4-carbonitrile;hydrochloride
SMILES C1CN(CCC1(C#N)C2=CC=CC=C2)CC3=CC=CC=C3.Cl

Specifications

Melting Point 263°C
Color White-Yellow
Quantity 5 g
UN Number 3439
Formula Weight 312.84
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzyl-4-cyano-4-phenylpiperidine Hydrochloride