missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-3-methylimidazolium chloride, 97%

Catalog No. AAH5999706
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5999706 5 g
AAH5999718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5999706 Supplier Thermo Scientific Chemicals Supplier No. H5999706

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 36443-80-8 | C11H13ClN2 | 208.689 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 36443-80-8
Molecular Formula C11H13ClN2
Molecular Weight (g/mol) 208.689
MDL Number MFCD07784451
InChI Key FCRZSGZCOGHOGF-UHFFFAOYSA-M
Synonym 1-benzyl-3-methylimidazolium chloride, 1-benzyl-3-methyl-1h-imidazol-3-ium chloride, 1h-imidazolium,1-methyl-3-phenylmethyl-, chloride 1:1, 3-benzyl-1-methylimidazol-1-ium chloride, dsstox_cid_27905, dsstox_rid_82658, dsstox_gsid_47929, ksc576i7l, 1-methyl-3-benzylimidazolium chloride, 1-benzyl-3-methylimidaz-olium chloride
PubChem CID 10560335
IUPAC Name 1-benzyl-3-methylimidazol-3-ium;chloride
SMILES C[N+]1=CN(C=C1)CC2=CC=CC=C2.[Cl-]

Specifications

Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 208.69
Percent Purity 97%
Chemical Name or Material 1-Benzyl-3-methylimidazolium chloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only