missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Benzyl-3-hydroxypiperidine 97.0+%, TCI America™
$165.90 - $497.70
Chemical Identifiers
| CAS | 14813-01-5 |
|---|---|
| Molecular Formula | C12H17NO |
| Molecular Weight (g/mol) | 191.27 |
| MDL Number | MFCD00023734 |
| InChI Key | UTTCOAGPVHRUFO-UHFFFAOYNA-N |
| Synonym | 1-benzyl-3-piperidinol, 1-benzyl-3-hydroxypiperidine, n-benzyl-3-hydroxypiperidine, 1-n-benzyl-3-hydroxy-piperidine, 3-piperidinol, 1-phenylmethyl, n-benzyl-3-piperidinol, pubchem7640, pubchem7727, acmc-209xh5, acmc-209xj8 |
| PubChem CID | 85773 |
| IUPAC Name | 1-benzylpiperidin-3-ol |
| SMILES | OC1CCCN(CC2=CC=CC=C2)C1 |
Chemical Identifiers
| 14813-01-5 | |
| 191.27 | |
| UTTCOAGPVHRUFO-UHFFFAOYNA-N | |
| 85773 | |
| OC1CCCN(CC2=CC=CC=C2)C1 |
| C12H17NO | |
| MFCD00023734 | |
| 1-benzyl-3-piperidinol, 1-benzyl-3-hydroxypiperidine, n-benzyl-3-hydroxypiperidine, 1-n-benzyl-3-hydroxy-piperidine, 3-piperidinol, 1-phenylmethyl, n-benzyl-3-piperidinol, pubchem7640, pubchem7727, acmc-209xh5, acmc-209xj8 | |
| 1-benzylpiperidin-3-ol |
Specifications
| 14813-01-5 | |
| 153°C | |
| MFCD00023734 | |
| 1-benzyl-3-piperidinol, 1-benzyl-3-hydroxypiperidine, n-benzyl-3-hydroxypiperidine, 1-n-benzyl-3-hydroxy-piperidine, 3-piperidinol, 1-phenylmethyl, n-benzyl-3-piperidinol, pubchem7640, pubchem7727, acmc-209xh5, acmc-209xj8 | |
| OC1CCCN(CC2=CC=CC=C2)C1 | |
| 191.27 | |
| 191.27 | |
| Liquid |
| Yellow | |
| C12H17NO | |
| 5 g | |
| UTTCOAGPVHRUFO-UHFFFAOYNA-N | |
| 1-benzylpiperidin-3-ol | |
| 85773 | |
| ≥97.0% (GC,T) | |
| 1-Benzyl-3-hydroxypiperidine |
Safety and Handling
TSCA : No