missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-2-pyrrolidinone, 97%

Catalog No. AC154120250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC154120250 25 g Glass bottle
AC154121000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC154120250 Supplier Thermo Scientific Chemicals Supplier No. 154120250

CAS: 5291-77-0 | C11H13NO | 175.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5291-77-0
Molecular Formula C11H13NO
Molecular Weight (g/mol) 175.23
MDL Number MFCD00003195
InChI Key LVUQCTGSDJLWCE-UHFFFAOYSA-N
Synonym 1-benzyl-2-pyrrolidinone, n-benzyl-2-pyrrolidinone, n-benzyl-2-pyrrolidone, 1-benzyl-2-pyrrolidone, n-benzylpyrrolidin-2-one, 2-pyrrolidinone, 1-phenylmethyl, n-benzylpyrrolidone, 1-benzylpyrrolidone, n-benzylpyrrolidinone, acmc-1aste
PubChem CID 79176
IUPAC Name 1-benzylpyrrolidin-2-one
SMILES C1CC(=O)N(C1)CC2=CC=CC=C2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0900g/mL
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.5500 to 1.553
Quantity 25 g
Specific Gravity 1.09
Formula Weight 175.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Benzyl-2-pyrrolidinone
Show More Show Less
EINECSNumber 226-131-5

RUO – Research Use Only