missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyl-2-methylimidazole 90.0+%, TCI America™

Catalog No. B0867100G
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0867100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B0867100G Supplier TCI America Supplier No. B0867100G
Only null left

Chemical Identifiers

CAS 13750-62-4
Molecular Formula C11H12N2
Molecular Weight (g/mol) 172.23
MDL Number MFCD00010637
InChI Key FBHPRUXJQNWTEW-UHFFFAOYSA-N
PubChem CID 83699
IUPAC Name 1-benzyl-2-methyl-1H-imidazole
SMILES CC1=NC=CN1CC1=CC=CC=C1

Specifications

Color White-Yellow
Boiling Point 127°C
Quantity 100 g
Formula Weight 172.23
Percent Purity ≥90.0% (GC)
Physical Form Wax
Chemical Name or Material 1-Benzyl-2-methylimidazole
EINECSNumber (5)-0416
RTECSNumber NI7722500
TSCA Yes