missing translation for 'onlineSavingsMsg'
Learn More

1-Benzoylpiperidine 98.0+%, TCI America™

Catalog No. B55125G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B55125G 5 g
B55121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B55125G Supplier TCI America Supplier No. B55125G

Chemical Identifiers

CAS 776-75-0
Molecular Formula C12H15NO
Molecular Weight (g/mol) 189.258
MDL Number MFCD00023702
InChI Key YXTROGRGRSPWKL-UHFFFAOYSA-N
Synonym 1-benzoylpiperidine, n-benzoylpiperidine, benzoylpiperidine, piperidine, 1-benzoyl, protectine i, phenyl piperidin-1-yl methanone, n-benzoylpiperidin, benzoic acid n-piperidide, alpha-repellin, benzoic acid, piperidide
PubChem CID 69892
IUPAC Name phenyl(piperidin-1-yl)methanone
SMILES C1CCN(CC1)C(=O)C2=CC=CC=C2

Specifications

Melting Point 50°C
Color White-Yellow
Boiling Point 321°C
Quantity 5 g
Formula Weight 189.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzoylpiperidine
RTECSNumber TM4600000
TSCA No