missing translation for 'onlineSavingsMsg'
Learn More

1-Benzoylpiperazine 98.0+%, TCI America™

Catalog No. B36765G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B36765G Supplier TCI America Supplier No. B36765G

Specifications

CAS 13754-38-6
Melting Point 68°C
Color White
Boiling Point 207°C
Molecular Formula C11H14N2O
Quantity 5 g
MDL Number MFCD00810192
Synonym 1-benzoylpiperazine, n-benzoylpiperazine, phenyl-piperazin-1-yl-methanone, 1-benzoyl piperazine, 1-benzoyl-piperazine, phenyl piperazin-1-yl methanone, piperazine, 1-benzoyl, phenyl piperazinyl ketone, phenyl piperazin-4-ium-1-yl methanone, 1-phenylcarbonyl piperazine
InChI Key VUNXBQRNMNVUMV-UHFFFAOYSA-N
SMILES C1CN(CCN1)C(=O)C2=CC=CC=C2
IUPAC Name phenyl(piperazin-1-yl)methanone
Molecular Weight (g/mol) 190.246
PubChem CID 762654
Formula Weight 190.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzoylpiperazine
Show More Show Less