missing translation for 'onlineSavingsMsg'
Learn More

1-Benzoylacetone, 98%, Thermo Scientific™

Chemical Identifiers

CAS 93-91-4
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00008786
InChI Key CVBUKMMMRLOKQR-UHFFFAOYSA-N
Synonym benzoylacetone, 1-phenyl-1,3-butanedione, 1-benzoylacetone, 1,3-butanedione, 1-phenyl, 1-benzoyl-2-propanone, acetoacetophenone, 2-propanone, benzoyl, acetylbenzoylmethane, 2-acetylacetophenone, benzoyl-aceton
PubChem CID 7166
IUPAC Name 1-phenylbutane-1,3-dione
SMILES CC(=O)CC(=O)C1=CC=CC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC105690500
SDS View Documents
Thermo Scientific Chemicals
105690500
50g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

93-91-4
162.19
CVBUKMMMRLOKQR-UHFFFAOYSA-N
7166
CC(=O)CC(=O)C1=CC=CC=C1
C10H10O2
MFCD00008786
benzoylacetone, 1-phenyl-1,3-butanedione, 1-benzoylacetone, 1,3-butanedione, 1-phenyl, 1-benzoyl-2-propanone, acetoacetophenone, 2-propanone, benzoyl, acetylbenzoylmethane, 2-acetylacetophenone, benzoyl-aceton
1-phenylbutane-1,3-dione

Specifications

93-91-4
100.0
Authentic
Glass bottle
C6H5COCH2COCH3
50g
1.09
Solubility in water: insoluble
CC(=O)CC(=O)C1=CC=CC=C1
162.19
162.19
1-Benzoylacetone, 98%
97.5
1.0900g/mL
98%
C10H10O2
MFCD00008786
07,68
benzoylacetone, 1-phenyl-1,3-butanedione, 1-benzoylacetone, 1,3-butanedione, 1-phenyl, 1-benzoyl-2-propanone, acetoacetophenone, 2-propanone, benzoyl, acetylbenzoylmethane, 2-acetylacetophenone, benzoyl-aceton
CVBUKMMMRLOKQR-UHFFFAOYSA-N
1-phenylbutane-1,3-dione
7166
98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 202-286-4

RTECSNumber : EK3540200

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only