missing translation for 'onlineSavingsMsg'
Learn More

1-Benzothiophene-2-sulfonyl chloride, 97%, Thermo Scientific™

Catalog No. CC12203CB
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC12203CB 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC12203CB Supplier Thermo Scientific Chemicals Supplier No. CC12203CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 90001-64-2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C8H5ClO2S2
Quantity 250 mg
MDL Number MFCD03659701
Synonym benzo b thiophene-2-sulfonyl chloride, 1-benzothiophene-2-sulfonylchloride, benzo b thiophene-2-sulfonylchloride, benzothiophene-2-sulfonyl chloride, pubchem10221, 2-benzothiophenesulfonyl chloride, benzo b thiophen-2-ylchlorosulfone, 1-benzothiophene-2-sulphonyl chloride, 2-chlorosulphonyl-1-benzothiophene, 1-benzo b thiophene-2-sulfonyl chloride
InChI Key FKIIVBOPAHICHQ-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C=C(S2)S(=O)(=O)Cl
IUPAC Name 1-benzothiophene-2-sulfonyl chloride
Molecular Weight (g/mol) 232.696
PubChem CID 2776333
Formula Weight 232.71
Percent Purity 97%
Chemical Name or Material 1-Benzothiophene-2-sulfonyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.