missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Benzothiophene-2-carbaldehyde, 97%, Thermo Scientific™
$817.25 - $817.25
Chemical Identifiers
| CAS | 3541-37-5 |
|---|---|
| Molecular Formula | C9H6OS |
| Molecular Weight (g/mol) | 162.21 |
| MDL Number | MFCD01075041 |
| InChI Key | NXSVNPSWARVMAY-UHFFFAOYSA-N |
| Synonym | benzo b thiophene-2-carboxaldehyde, benzo b thiophene-2-carbaldehyde, thianaphthene-2-carboxaldehyde, benzothiophene-2-carboxaldehyde, acmc-209igo, 2-formylbenzo b thiophene, 2-benzothiophenecarbaldehyde, 2-formyl-benzo b thiophene, benzothiophene-2-carbaldehyde, 2-benzothiophenecarboxaldehyde |
| PubChem CID | 736500 |
| IUPAC Name | 1-benzothiophene-2-carbaldehyde |
| SMILES | O=CC1=CC2=CC=CC=C2S1 |
Chemical Identifiers
| 3541-37-5 | |
| 162.21 | |
| NXSVNPSWARVMAY-UHFFFAOYSA-N | |
| 736500 | |
| O=CC1=CC2=CC=CC=C2S1 |
| C9H6OS | |
| MFCD01075041 | |
| benzo b thiophene-2-carboxaldehyde, benzo b thiophene-2-carbaldehyde, thianaphthene-2-carboxaldehyde, benzothiophene-2-carboxaldehyde, acmc-209igo, 2-formylbenzo b thiophene, 2-benzothiophenecarbaldehyde, 2-formyl-benzo b thiophene, benzothiophene-2-carbaldehyde, 2-benzothiophenecarboxaldehyde | |
| 1-benzothiophene-2-carbaldehyde |
Specifications
| 3541-37-5 | |
| 100.0 | |
| Amber Glass Bottle | |
| Yes | |
| 10 g | |
| NXSVNPSWARVMAY-UHFFFAOYSA-N | |
| 1-benzothiophene-2-carbaldehyde | |
| 736500 | |
| 97% |
| 96.5 | |
| 97% | |
| C9H6OS | |
| MFCD01075041 | |
| benzo b thiophene-2-carboxaldehyde, benzo b thiophene-2-carbaldehyde, thianaphthene-2-carboxaldehyde, benzothiophene-2-carboxaldehyde, acmc-209igo, 2-formylbenzo b thiophene, 2-benzothiophenecarbaldehyde, 2-formyl-benzo b thiophene, benzothiophene-2-carbaldehyde, 2-benzothiophenecarboxaldehyde | |
| O=CC1=CC2=CC=CC=C2S1 | |
| 162.21 | |
| 162.21 | |
| 1-Benzothiophene-2-carbaldehyde |
Safety and Handling
GHS P Statement Causes serious eye irritation.
Warning