missing translation for 'onlineSavingsMsg'
Learn More

1-Benzothiophen-3-ylmethylamine hydrochloride, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC12313DE

 View more versions of this product

Catalog No. CC12313DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

1-Benzothiophen-3-ylmethylamine hydrochloride
80.0
95%
MFCD01566652
benzo b thiophen-3-ylmethanamine hydrochloride, 1-benzothiophen-3-ylmethanamine hydrochloride, 1-benzothiophen-3-ylmethylamine hydrochloride, benzo b thiophen-3-ylmethylamine hydrochloride, 1-1-benzothiophen-3-yl methanamine hydrochloride, benzo b thiophen-3-ylmethylamine, chloride, 1-benzothiophen-3-yl methanamine hydrochloride, 3-aminomethyl benzo b thiophene hydrochloride, benzo b thiophen-3-yl-methylamine hydrochloride salt, 1-1-benzothiophen-3-yl methanamine-hydrogen chloride 1/1
C1=CC=C2C(=C1)C(=CS2)CN.Cl
199.696
199.7
55810-74-7
100.0
C9H10ClNS
5g
MIAPJPCDRSPNPW-UHFFFAOYSA-N
1-benzothiophen-3-ylmethanamine;hydrochloride
2776346
95%

Chemical Identifiers

55810-74-7
199.696
MIAPJPCDRSPNPW-UHFFFAOYSA-N
2776346
C1=CC=C2C(=C1)C(=CS2)CN.Cl
C9H10ClNS
MFCD01566652
benzo b thiophen-3-ylmethanamine hydrochloride, 1-benzothiophen-3-ylmethanamine hydrochloride, 1-benzothiophen-3-ylmethylamine hydrochloride, benzo b thiophen-3-ylmethylamine hydrochloride, 1-1-benzothiophen-3-yl methanamine hydrochloride, benzo b thiophen-3-ylmethylamine, chloride, 1-benzothiophen-3-yl methanamine hydrochloride, 3-aminomethyl benzo b thiophene hydrochloride, benzo b thiophen-3-yl-methylamine hydrochloride salt, 1-1-benzothiophen-3-yl methanamine-hydrogen chloride 1/1
1-benzothiophen-3-ylmethanamine;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions