Learn More
1-Benzothiophen-3-ylmethylamine hydrochloride, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC12313DA
Specifications
| 1-Benzothiophen-3-ylmethylamine hydrochloride | |
| 80.0 | |
| 95% | |
| C9H10ClNS | |
| 1g | |
| MIAPJPCDRSPNPW-UHFFFAOYSA-N | |
| 1-benzothiophen-3-ylmethanamine;hydrochloride | |
| 2776346 | |
| 95% |
| 55810-74-7 | |
| 100.0 | |
| Amber glass bottle | |
| MFCD01566652 | |
| benzo b thiophen-3-ylmethanamine hydrochloride, 1-benzothiophen-3-ylmethanamine hydrochloride, 1-benzothiophen-3-ylmethylamine hydrochloride, benzo b thiophen-3-ylmethylamine hydrochloride, 1-1-benzothiophen-3-yl methanamine hydrochloride, benzo b thiophen-3-ylmethylamine, chloride, 1-benzothiophen-3-yl methanamine hydrochloride, 3-aminomethyl benzo b thiophene hydrochloride, benzo b thiophen-3-yl-methylamine hydrochloride salt, 1-1-benzothiophen-3-yl methanamine-hydrogen chloride 1/1 | |
| C1=CC=C2C(=C1)C(=CS2)CN.Cl | |
| 199.696 | |
| 199.7 |
Chemical Identifiers
| 55810-74-7 | |
| 199.696 | |
| MIAPJPCDRSPNPW-UHFFFAOYSA-N | |
| 2776346 | |
| C1=CC=C2C(=C1)C(=CS2)CN.Cl |
| C9H10ClNS | |
| MFCD01566652 | |
| benzo b thiophen-3-ylmethanamine hydrochloride, 1-benzothiophen-3-ylmethanamine hydrochloride, 1-benzothiophen-3-ylmethylamine hydrochloride, benzo b thiophen-3-ylmethylamine hydrochloride, 1-1-benzothiophen-3-yl methanamine hydrochloride, benzo b thiophen-3-ylmethylamine, chloride, 1-benzothiophen-3-yl methanamine hydrochloride, 3-aminomethyl benzo b thiophene hydrochloride, benzo b thiophen-3-yl-methylamine hydrochloride salt, 1-1-benzothiophen-3-yl methanamine-hydrogen chloride 1/1 | |
| 1-benzothiophen-3-ylmethanamine;hydrochloride |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning