missing translation for 'onlineSavingsMsg'
Learn More

1-Benzothiophen-2-ylmethylamine, 95%, Thermo Scientific™

Catalog No. CC12213DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC12213DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC12213DA Supplier Thermo Scientific Chemicals Supplier No. CC12213DA

Chemical Identifiers

CAS 6314-43-8
Molecular Formula C9H9NS
Molecular Weight (g/mol) 163.238
MDL Number MFCD00965305
InChI Key WKYFWFHTABURGB-UHFFFAOYSA-N
Synonym benzo b thiophen-2-ylmethanamine, 1-benzothiophen-2-ylmethylamine, benzo b thiophene-2-methanamine, c-benzo b thiophen-2-yl-methylamine, benzo b thiophen-2-ylmethylamine, 2-aminomethyl benzo b thiophene, 1-benzothiophene-2-yl methylamine, 1-1-benzothiophen-2-yl methanamine, benzo b thiophene-2-methylamine
PubChem CID 237074
IUPAC Name 1-benzothiophen-2-ylmethanamine
SMILES C1=CC=C2C(=C1)C=C(S2)CN

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 163.24
Percent Purity 95%
Chemical Name or Material 1-Benzothiophen-2-ylmethylamine
Hazard Category Danger
Hazard Statement GHS P Statement Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Co