missing translation for 'onlineSavingsMsg'
Learn More

1-Benzofuran-5-carboxylic acid, Thermo Scientific™

Catalog No. p-4509659
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC07101DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC07101DA Supplier Thermo Scientific Chemicals Supplier No. CC07101DA

Chemical Identifiers

CAS 90721-27-0
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.14
MDL Number MFCD01006742
InChI Key GTWXSZIQNTUNKR-UHFFFAOYSA-N
Synonym benzofuran-5-carboxylic acid, 5-benzofurancarboxylic acid, benzo b furan-5-carboxylic acid, 1-benzofunan-5-carboxylic acid, benzofurane-5-carboxylic acid, 4ctj, pubchem7018, 5-benzofuran carboxylic, rarechem al be 1369, 5-benzofurancarboxylicacid
PubChem CID 595656
IUPAC Name 1-benzofuran-5-carboxylic acid
SMILES OC(=O)C1=CC=C2OC=CC2=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 162.14
Percent Purity 97%
Chemical Name or Material 1-Benzofuran-5-carboxylic acid
Hazard Category Warning