Learn More
1-Benzofuran-5-carbaldehyde, 97%, Thermo Scientific™
$149.00 - $149.00
Chemical Identifiers
CAS | 10035-16-2 |
---|---|
Molecular Formula | C9H6O2 |
Molecular Weight (g/mol) | 146.145 |
MDL Number | MFCD03411182 |
InChI Key | LLLBDLDNTMMZHL-UHFFFAOYSA-N |
Synonym | benzofuran-5-carbaldehyde, 5-formylbenzofuran, 5-benzofurancarboxaldehyde, benzofuran-5-carboxaldehyde, 5-formyl-1-benzofuran, 5-formylbenzo b furan, benzo b furan-5-carboxaldehyde, 1-benzofuran-5-carboxaldehyde, benzo b furan-5-carbaldehyde |
PubChem CID | 2773875 |
IUPAC Name | 1-benzofuran-5-carbaldehyde |
SMILES | C1=CC2=C(C=CO2)C=C1C=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
CC07104CB
|
Thermo Scientific Chemicals
CC07104CB |
250 mg | Amber Glass Bottle |
Each for $149.00
|
|
CC07104DE
|
Thermo Scientific Chemicals
CC07104DE |
5g | Amber Glass Bottle |
N/A
|
N/A |
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
10035-16-2 | |
146.145 | |
LLLBDLDNTMMZHL-UHFFFAOYSA-N | |
2773875 | |
C1=CC2=C(C=CO2)C=C1C=O |
C9H6O2 | |
MFCD03411182 | |
benzofuran-5-carbaldehyde, 5-formylbenzofuran, 5-benzofurancarboxaldehyde, benzofuran-5-carboxaldehyde, 5-formyl-1-benzofuran, 5-formylbenzo b furan, benzo b furan-5-carboxaldehyde, 1-benzofuran-5-carboxaldehyde, benzo b furan-5-carbaldehyde | |
1-benzofuran-5-carbaldehyde |
Specifications
10035-16-2 | |
100.0 | |
C9H6O2 | |
Amber Glass Bottle | |
benzofuran-5-carbaldehyde, 5-formylbenzofuran, 5-benzofurancarboxaldehyde, benzofuran-5-carboxaldehyde, 5-formyl-1-benzofuran, 5-formylbenzo b furan, benzo b furan-5-carboxaldehyde, 1-benzofuran-5-carboxaldehyde, benzo b furan-5-carbaldehyde | |
C1=CC2=C(C=CO2)C=C1C=O | |
146.145 | |
146.14 | |
1-Benzofuran-5-carbaldehyde |
96.5 | |
97% | |
MFCD03411182 | |
250 mg | |
LLLBDLDNTMMZHL-UHFFFAOYSA-N | |
1-benzofuran-5-carbaldehyde | |
2773875 | |
97% |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.