missing translation for 'onlineSavingsMsg'
Learn More

1-benzofuran-5-amine, 97%, Thermo Scientific™

Catalog No. CC07114DE
Change view
Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC07114DE 5g Amber Glass Bottle
CC07114CB 250mg Amber Glass Bottle
CC07114DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC07114DE Supplier Thermo Scientific Chemicals Supplier No. CC07114DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 58546-89-7
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
InChI Key GMOLCSICTCPZCU-UHFFFAOYSA-N
Synonym benzofuran-5-amine, 5-benzofuranamine, 5-aminobenzo b furan, 5-aminobenzofuran, benzo b furan-5-amine, benzo b furan-5-ylamine, benzofuran-5-ylamine
PubChem CID 1477152
IUPAC Name 1-benzofuran-5-amine
SMILES C1=CC2=C(C=CO2)C=C1N

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5g
Formula Weight 133.15
Percent Purity 97%
Chemical Name or Material 1-Benzofuran-5-amine