missing translation for 'onlineSavingsMsg'
Learn More

1-benzofuran-5-amine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC07114DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC07114DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

58546-89-7
133.15
benzofuran-5-amine, 5-benzofuranamine, 5-aminobenzo b furan, 5-aminobenzofuran, benzo b furan-5-amine, benzo b furan-5-ylamine, benzofuran-5-ylamine
1-benzofuran-5-amine
C8H7NO
GMOLCSICTCPZCU-UHFFFAOYSA-N
1477152
C1=CC2=C(C=CO2)C=C1N

Specifications

1-Benzofuran-5-amine
97%
C8H7NO
benzofuran-5-amine, 5-benzofuranamine, 5-aminobenzo b furan, 5-aminobenzofuran, benzo b furan-5-amine, benzo b furan-5-ylamine, benzofuran-5-ylamine
C1=CC2=C(C=CO2)C=C1N
133.15
133.15
58546-89-7
Amber Glass Bottle
1g
GMOLCSICTCPZCU-UHFFFAOYSA-N
1-benzofuran-5-amine
1477152
97%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions