missing translation for 'onlineSavingsMsg'
Learn More

1-Benzofuran-2-sulfonyl chloride, Thermo Scientific™

Catalog No. CC06603CB
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC06603CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC06603CB Supplier Thermo Scientific Chemicals Supplier No. CC06603CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17070-58-5
Molecular Formula C8H5ClO3S
Molecular Weight (g/mol) 216.635
InChI Key RWCKBBSBRTUUHR-UHFFFAOYSA-N
Synonym benzofuran-2-sulfonyl chloride, 2-benzofuransulfonylchloride, 2-benzofuransulfonyl chloride, benzo b furan-2-sulfonyl chloride, pubchem7024, benzo d furan-2-ylchlorosulfone
PubChem CID 2795172
IUPAC Name 1-benzofuran-2-sulfonyl chloride
SMILES C1=CC=C2C(=C1)C=C(O2)S(=O)(=O)Cl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 216.64
Percent Purity 97%
Chemical Name or Material 1-Benzofuran-2-sulfonyl chloride