missing translation for 'onlineSavingsMsg'
Learn More

1-Benzo[b]thiophen-3-yl-2-bromoethan-1-one, 97%, Thermo Scientific™

Catalog No. CC12305DA
Change view
Click to view available options
Quantity:
1g
5g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC12305DA 1g Amber Glass Bottle
CC12305DE 5g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC12305DA Supplier Thermo Scientific Chemicals Supplier No. CC12305DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 26167-45-3
Molecular Formula C10H7BrOS
Molecular Weight (g/mol) 255.129
InChI Key CKHWNGWAHFLCTJ-UHFFFAOYSA-N
Synonym 1-benzo b thiophen-3-yl-2-bromoethanone, 1-benzo b thiophen-3-yl-2-bromoethan-1-one, 3-bromoacetyl benzothiophene, 1-1-benzothiophen-3-yl-2-bromoethan-1-one, 3-bromoacetyl benzo b thiophene, 1-1-benzothiophen-3-yl-2-bromoethanone, 1-1-benzothiophen-3-yl-2-bromo-1-ethanone, 3-2-bromoacetyl benzo b thiophen, 1-benzothiophen-3-yl-2-bromo-ethanone, ethanone,1-benzo b thien-3-yl-2-bromo
PubChem CID 2776341
IUPAC Name 1-(1-benzothiophen-3-yl)-2-bromoethanone
SMILES C1=CC=C2C(=C1)C(=CS2)C(=O)CBr

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 255.13
Percent Purity 97%
Chemical Name or Material 1-Benzo[b]thiophen-3-yl-2-bromoethan-1-one