missing translation for 'onlineSavingsMsg'
Learn More

1-Aza-18-crown 6-Ether 98.0+%, TCI America™

Catalog No. A13245G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13245G 5 g
A13241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A13245G Supplier TCI America Supplier No. A13245G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 33941-15-0
Molecular Formula C12H25NO5
Molecular Weight (g/mol) 263.334
MDL Number MFCD00075466
InChI Key NBXKUSNBCPPKRA-UHFFFAOYSA-N
PubChem CID 118578
IUPAC Name 1,4,7,10,13-pentaoxa-16-azacyclooctadecane
SMILES C1COCCOCCOCCOCCOCCN1

Specifications

Melting Point 51°C
Quantity 5 g
Synonym 1-aza-18-crown-6, 1-aza-18-crown6-ether, aza-18-crown-6, acmc-209tw0, 1-aza-18-crown 6-ether, 1-aza-18-crown nt, 1-aza-4,7,10,13,16-pentaoxacyclooctadecane
Formula Weight 263.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Aza-18-crown 6-Ether