missing translation for 'onlineSavingsMsg'
Learn More

1-Aminoundecane 98.0+%, TCI America™

Catalog No. A076110ML
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A076110ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A076110ML Supplier TCI America Supplier No. A076110ML
Only null left

Chemical Identifiers

CAS 7307-55-3
Molecular Formula C11H25N
Molecular Weight (g/mol) 171.33
MDL Number MFCD00008152
InChI Key QFKMMXYLAPZKIB-UHFFFAOYSA-N
Synonym undecylamine, n-undecylamine, 1-aminoundecane, 1-undecanamine, hendecylamine, 1-undecylamine, monoundecylamine, n-undecylamin, unii-uze26a4ko1, uze26a4ko1
PubChem CID 81742
IUPAC Name undecan-1-amine
SMILES CCCCCCCCCCCN

Specifications

Melting Point 17°C
Color Colorless
Boiling Point 242°C
Quantity 10 mL
UN Number 2735
Formula Weight 171.33
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Aminoundecane
EINECSNumber (2)-0133
RTECSNumber YQ1518000
TSCA No