missing translation for 'onlineSavingsMsg'
Learn More

1-Aminopyridinium Iodide 98.0+%, TCI America™

Catalog No. p-7116974
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
A06295G 5 g
A062925G 25 g
2 options

Catalog No. A06295G

Supplier: TCI America A06295G

Only null left

Chemical Identifiers

CAS 6295-87-0
Molecular Formula C5H7IN2
Molecular Weight (g/mol) 222.03
MDL Number MFCD00051893
InChI Key NDRLPYIMWROJBG-UHFFFAOYSA-M
Synonym 1-aminopyridinium iodide, 1-aminopyridin-1-ium iodide, n-aminopyridinium iodide, pyridinium, 1-amino-, iodide, 1-aminopyridiniumiodide, pyridylamine, iodide, aminopyridinium iodide, amino-pyridinium iodide, l-aminopyridinium iodide, acmc-209nay
PubChem CID 2733704
IUPAC Name 1-aminopyridin-1-ium iodide
SMILES [I-].N[N+]1=CC=CC=C1

Specifications

Quantity 5 g
Formula Weight 222.03
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Aminopyridinium Iodide