missing translation for 'onlineSavingsMsg'
Learn More

1-Aminopyridinium iodide, 97%

Catalog No. AAB2570314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2570314 25 g
AAB2570306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2570314 Supplier Thermo Scientific Chemicals Supplier No. B2570314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6295-87-0 | C5H7IN2 | 222.03 g/mol

1-Aminopyridinium iodide is a reagent used as a building block to construct fused heterocycles, synthesis of substituted pyridines, dipolar cycloadditions, ylide type reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Aminopyridinium iodide is a reagent used as a building block to construct fused heterocycles, synthesis of substituted pyridines, dipolar cycloadditions, ylide type reactions.

Solubility
Soluble in water (almost transparency).

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 6295-87-0
Molecular Formula C5H7IN2
Molecular Weight (g/mol) 222.03
MDL Number MFCD00051893
InChI Key NDRLPYIMWROJBG-UHFFFAOYSA-M
Synonym 1-aminopyridinium iodide, 1-aminopyridin-1-ium iodide, n-aminopyridinium iodide, pyridinium, 1-amino-, iodide, 1-aminopyridiniumiodide, pyridylamine, iodide, aminopyridinium iodide, amino-pyridinium iodide, l-aminopyridinium iodide, acmc-209nay
PubChem CID 2733704
IUPAC Name pyridin-1-ium-1-amine;iodide
SMILES [I-].N[N+]1=CC=CC=C1

Specifications

Melting Point ∼160°C (decomposition)
Quantity 25 g
Beilstein 3713241
Solubility Information Soluble in water (almost transparency).
Formula Weight 222.03
Percent Purity 97%
Chemical Name or Material 1-Aminopyridinium iodide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 228-566-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only