missing translation for 'onlineSavingsMsg'
Learn More

1-Aminopiperidine, 97%

Catalog No. p-3211366
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC104410100 10 g Glass bottle
AC104410250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC104410100 Supplier Thermo Scientific Chemicals Supplier No. 104410100
Only null left

CAS: 2213-43-6 | C5H12N2 | 100.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2213-43-6
Molecular Formula C5H12N2
Molecular Weight (g/mol) 100.16
MDL Number MFCD00006489
InChI Key LWMPFIOTEAXAGV-UHFFFAOYSA-N
Synonym 1-aminopiperidine, n-aminopiperidine, 1-piperidinamine, piperidine, 1-amino, n-piperidylamine, aminopiperidine, piperidinamine, 1,1-pentamethylenehydrazine, piperidylamine, piperidine amine
PubChem CID 16658
IUPAC Name piperidin-1-amine
SMILES C1CCN(CC1)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Gray or White to Yellow
Density 0.9280g/mL
Boiling Point 145.0°C to 147.0°C
Flash Point 36°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4750 to 1.4770
Quantity 10 g
Beilstein 20, 89
Specific Gravity 0.928
Solubility Information Solubility in water: miscible with water. Other solubilities: miscible with toluene and alcohol
Formula Weight 100.16
Percent Purity 97%
Physical Form Crystals or Powder
Chemical Name or Material 1-Aminopiperidine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.
Harmful if inhaled.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 218-666-8

RUO – Research Use Only