Learn More
1-Aminopiperidine, 97%
CAS: 2213-43-6 | C5H12N2 | 100.165 g/mol
Supplier: Thermo Scientific Chemicals L1407822
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1-Aminopiperidine | |
| 0.928 | |
| 36°C (96°F) | |
| 1.475 | |
| 100 g | |
| 383565 | |
| LWMPFIOTEAXAGV-UHFFFAOYSA-N | |
| piperidin-1-amine | |
| 16658 | |
| 97% |
| 2213-43-6 | |
| 145°C to 147°C | |
| C5H12N2 | |
| MFCD00006489 | |
| UN2733 | |
| 1-aminopiperidine, n-aminopiperidine, 1-piperidinamine, piperidine, 1-amino, n-piperidylamine, aminopiperidine, piperidinamine, 1,1-pentamethylenehydrazine, piperidylamine, piperidine amine | |
| C1CCN(CC1)N | |
| 100.165 | |
| 100.16 |
Chemical Identifiers
| 2213-43-6 | |
| 100.165 | |
| LWMPFIOTEAXAGV-UHFFFAOYSA-N | |
| 16658 | |
| C1CCN(CC1)N |
| C5H12N2 | |
| MFCD00006489 | |
| 1-aminopiperidine, n-aminopiperidine, 1-piperidinamine, piperidine, 1-amino, n-piperidylamine, aminopiperidine, piperidinamine, 1,1-pentamethylenehydrazine, piperidylamine, piperidine amine | |
| piperidin-1-amine |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H226-H302+H312+H332-H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 218-666-8
RTECSNumber : TM4165000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only