missing translation for 'onlineSavingsMsg'
Learn More

1-Aminopiperidine, 97%

Catalog No. AAL1407806
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1407806 5 g
AAL1407814 25 g
AAL1407822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1407806 Supplier Thermo Scientific Chemicals Supplier No. L1407806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2213-43-6 | C5H12N2 | 100.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2213-43-6
Molecular Formula C5H12N2
Molecular Weight (g/mol) 100.165
MDL Number MFCD00006489
InChI Key LWMPFIOTEAXAGV-UHFFFAOYSA-N
Synonym 1-aminopiperidine, n-aminopiperidine, 1-piperidinamine, piperidine, 1-amino, n-piperidylamine, aminopiperidine, piperidinamine, 1,1-pentamethylenehydrazine, piperidylamine, piperidine amine
PubChem CID 16658
IUPAC Name piperidin-1-amine
SMILES C1CCN(CC1)N

Specifications

Density 0.928
Boiling Point 145°C to 147°C
Flash Point 36°C (96°F)
Refractive Index 1.475
Quantity 5 g
UN Number UN2733
Beilstein 383565
Formula Weight 100.16
Percent Purity 97%
Chemical Name or Material 1-Aminopiperidine
Hazard Category H226-H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 218-666-8
RTECSNumber TM4165000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only