missing translation for 'onlineSavingsMsg'
Learn More

1-Aminononadecane 96.0+%, TCI America™

Catalog No. A076510G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A076510G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A076510G Supplier TCI America Supplier No. A076510G
Only null left

Chemical Identifiers

CAS 14130-05-3
Molecular Formula C19H41N
Molecular Weight (g/mol) 283.54
MDL Number MFCD00047944
InChI Key INAMEDPXUAWNKL-UHFFFAOYSA-N
Synonym Nonadecylamine, Nonadecane-1-amine
PubChem CID 106416
IUPAC Name nonadecan-1-amine
SMILES CCCCCCCCCCCCCCCCCCCN

Specifications

Melting Point 53°C
Color White
Quantity 10 g
Formula Weight 283.54
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Aminononadecane
EINECSNumber (2)-0133
TSCA No
Recommended Storage Refrigerator