missing translation for 'onlineSavingsMsg'
Learn More

1-Aminohydantoin hydrochloride, 98%

CAS: 2827-56-7 | C3H5N3O2·ClH | 151.55 g/mol

$115.71 - $115.71

Chemical Identifiers

CAS 2827-56-7
Molecular Formula C3H5N3O2·ClH
Molecular Weight (g/mol) 151.55
InChI Key WEOHANUVLKERQI-UHFFFAOYSA-N
Synonym 1-aminohydantoin hydrochloride, 1-aminoimidazolidine-2,4-dione hydrochloride, 1-aminohydantoin hcl, 1-amino hydantoin hydrochloride, 2,4-imidazolidinedione, 1-amino-, monohydrochloride, acmc-209h1m, 1-aminoimidazolidine-2,4-dione hcl, 1-aminoimidazoline-2,4-dione hydrochloride
PubChem CID 12472963
IUPAC Name 1-aminoimidazolidine-2,4-dione;hydrochloride
SMILES C1C(=O)NC(=O)N1N.Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC442050050
SDS View Documents
Thermo Scientific Chemicals
442050050
5 g Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2827-56-7
151.55
1-aminohydantoin hydrochloride, 1-aminoimidazolidine-2,4-dione hydrochloride, 1-aminohydantoin hcl, 1-amino hydantoin hydrochloride, 2,4-imidazolidinedione, 1-amino-, monohydrochloride, acmc-209h1m, 1-aminoimidazolidine-2,4-dione hcl, 1-aminoimidazoline-2,4-dione hydrochloride
1-aminoimidazolidine-2,4-dione;hydrochloride
C3H5N3O2·ClH
WEOHANUVLKERQI-UHFFFAOYSA-N
12472963
C1C(=O)NC(=O)N1N.Cl

Specifications

2827-56-7
100.0
Yellow
97.5 to 102.5% (Argentometry)
C3H5N3O2·ClH
1-aminohydantoin hydrochloride, 1-aminoimidazolidine-2,4-dione hydrochloride, 1-aminohydantoin hcl, 1-amino hydantoin hydrochloride, 2,4-imidazolidinedione, 1-amino-, monohydrochloride, acmc-209h1m, 1-aminoimidazolidine-2,4-dione hcl, 1-aminoimidazoline-2,4-dione hydrochloride
C1C(=O)NC(=O)N1N.Cl
151.55
151.55
Crystalline Powder
97.5
201.0°C to 205.0°C
Authentic
Glass bottle
5 g
WEOHANUVLKERQI-UHFFFAOYSA-N
1-aminoimidazolidine-2,4-dione;hydrochloride
12472963
98%
1-Aminohydantoin hydrochloride
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only