missing translation for 'onlineSavingsMsg'
Learn More

1-Aminohydantoin hydrochloride, 90%

Catalog No. AC442050050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC442050050 5 g Glass bottle
AC442050250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC442050050 Supplier Thermo Scientific Chemicals Supplier No. 442050050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2827-56-7 | C3H5N3O2·ClH | 151.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2827-56-7
Molecular Formula C3H5N3O2·ClH
Molecular Weight (g/mol) 151.55
InChI Key WEOHANUVLKERQI-UHFFFAOYSA-N
Synonym 1-aminohydantoin hydrochloride, 1-aminoimidazolidine-2,4-dione hydrochloride, 1-aminohydantoin hcl, 1-amino hydantoin hydrochloride, 2,4-imidazolidinedione, 1-amino-, monohydrochloride, acmc-209h1m, 1-aminoimidazolidine-2,4-dione hcl, 1-aminoimidazoline-2,4-dione hydrochloride
PubChem CID 12472963
IUPAC Name 1-aminoimidazolidine-2,4-dione;hydrochloride
SMILES C1C(=O)NC(=O)N1N.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 201.0°C to 205.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5 to 102.5% (Argentometry)
Packaging Glass bottle
Quantity 5 g
Formula Weight 151.55
Percent Purity 90%
Physical Form Crystalline Powder
Chemical Name or Material 1-Aminohydantoin hydrochloride
Show More Show Less

RUO – Research Use Only