missing translation for 'onlineSavingsMsg'
Learn More

1-Aminodecane 98.0+%, TCI America™

Catalog No. p-7125994
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D003325ML 25 mL
D0033100ML 100 mL
D0033500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D003325ML Supplier TCI America Supplier No. D003325ML
Only null left

Chemical Identifiers

CAS 2016-57-1
Molecular Formula C10H23N
Molecular Weight (g/mol) 157.30
MDL Number MFCD00008149
InChI Key MHZGKXUYDGKKIU-UHFFFAOYSA-N
Synonym decylamine, 1-aminodecane, 1-decanamine, n-decylamine, 1-decylamine, decanamine, monodecylamine, kemamine p 190d, aminodecane, decyl amine
PubChem CID 8916
IUPAC Name decan-1-amine
SMILES CCCCCCCCCCN

Specifications

Color Yellow
Boiling Point 220°C
Quantity 25 mL
UN Number 2922
Formula Weight 157.30
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 1-Aminodecane
EINECSNumber (2)-0133
RTECSNumber HD6475000
TSCA Yes