missing translation for 'onlineSavingsMsg'
Learn More

1-Aminodécane 98,0+%, TCI America™

Numéro de catalogue. D0033500ML
Modifier l'affichage
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
D0033500ML 500 mL
D003325ML 25 mL
D0033100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. D0033500ML Fournisseur TCI America Code fournisseur D0033500ML

Identifiants chimiques

CAS 2016-57-1
Molecular Formula C10H23N
Molecular Weight (g/mol) 157.30
MDL Number MFCD00008149
InChI Key MHZGKXUYDGKKIU-UHFFFAOYSA-N
Synonym decylamine, 1-aminodecane, 1-decanamine, n-decylamine, 1-decylamine, decanamine, monodecylamine, kemamine p 190d, aminodecane, decyl amine
PubChem CID 8916
IUPAC Name Decan-1-amine
SMILES CCCCCCCCCCN

Spécifications

Color Jaune
Boiling Point 220°C
Quantity 500 mL
UN Number 2922
Formula Weight 157.30
Percent Purity ≥98.0% (T)
Physical Form Liquide
Chemical Name or Material 1-Aminodecane
EINECSNumber (2)-0133
RTECSNumber HD6475000
TSCA Yes