missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Aminocyclopropanecarboxylic Acid 97.0+%, TCI America™
$33.56 - $331.81
Chemical Identifiers
| CAS | 22059-21-8 |
|---|---|
| Molecular Formula | C4H7NO2 |
| Molecular Weight (g/mol) | 101.11 |
| MDL Number | MFCD00009944 |
| InChI Key | PAJPWUMXBYXFCZ-UHFFFAOYSA-N |
| Synonym | 1-aminocyclopropanecarboxylic acid, 1-amino-1-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-amino, carboxycyclopropylamine, acc, alpha-aminocyclopropanecarboxylic acid, alpha-aminocyclopropane carboxylic acid, 1-amino-cyclopropanecarboxylic acid, h-acpc-oh, aminocyclopropanecarboxylic acid |
| PubChem CID | 535 |
| ChEBI | CHEBI:18053 |
| IUPAC Name | 1-aminocyclopropane-1-carboxylic acid |
| SMILES | NC1(CC1)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A1178100MG
|
TCI America
A1178100MG |
100 mg |
Each for $33.56
|
|
|||||
|
A11781G
|
TCI America
A11781G |
1 g |
Each for $141.67
|
|
|||||
|
A11785G
|
TCI America
A11785G |
5 g |
Each for $331.81
|
|
|||||
Chemical Identifiers
| 22059-21-8 | |
| 101.11 | |
| PAJPWUMXBYXFCZ-UHFFFAOYSA-N | |
| 535 | |
| 1-aminocyclopropane-1-carboxylic acid |
| C4H7NO2 | |
| MFCD00009944 | |
| 1-aminocyclopropanecarboxylic acid, 1-amino-1-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-amino, carboxycyclopropylamine, acc, alpha-aminocyclopropanecarboxylic acid, alpha-aminocyclopropane carboxylic acid, 1-amino-cyclopropanecarboxylic acid, h-acpc-oh, aminocyclopropanecarboxylic acid | |
| CHEBI:18053 | |
| NC1(CC1)C(O)=O |
Specifications
| 22059-21-8 | |
| White | |
| MFCD00009944 | |
| 1-aminocyclopropanecarboxylic acid, 1-amino-1-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-amino, carboxycyclopropylamine, acc, alpha-aminocyclopropanecarboxylic acid, alpha-aminocyclopropane carboxylic acid, 1-amino-cyclopropanecarboxylic acid, h-acpc-oh, aminocyclopropanecarboxylic acid | |
| NC1(CC1)C(O)=O | |
| 101.11 | |
| CHEBI:18053 | |
| ≥97.0% (T) | |
| 1-Aminocyclopropanecarboxylic Acid |
| 231°C | |
| C4H7NO2 | |
| 100 mg | |
| PAJPWUMXBYXFCZ-UHFFFAOYSA-N | |
| 1-aminocyclopropane-1-carboxylic acid | |
| 535 | |
| 101.11 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : GZ1110000
TSCA : No