missing translation for 'onlineSavingsMsg'
Learn More

1-Aminocyclopropanecarboxylic Acid 97.0+%, TCI America™

Catalog No. A1178100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1178100MG 100 mg
A11781G 1 g
A11785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A1178100MG Supplier TCI America Supplier No. A1178100MG

Chemical Identifiers

CAS 22059-21-8
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.11
MDL Number MFCD00009944
InChI Key PAJPWUMXBYXFCZ-UHFFFAOYSA-N
Synonym 1-aminocyclopropanecarboxylic acid, 1-amino-1-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 1-amino, carboxycyclopropylamine, acc, alpha-aminocyclopropanecarboxylic acid, alpha-aminocyclopropane carboxylic acid, 1-amino-cyclopropanecarboxylic acid, h-acpc-oh, aminocyclopropanecarboxylic acid
PubChem CID 535
ChEBI CHEBI:18053
IUPAC Name 1-aminocyclopropane-1-carboxylic acid
SMILES NC1(CC1)C(O)=O

Specifications

Melting Point 231°C
Color White
Quantity 100 mg
Formula Weight 101.11
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Aminocyclopropanecarboxylic Acid
RTECSNumber GZ1110000
TSCA No