missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Aminocyclohexanecarboxylic Acid 98.0+%, TCI America™
$334.98 - $1197.46
Chemical Identifiers
| CAS | 2756-85-6 |
|---|---|
| Molecular Formula | C7H13NO2 |
| Molecular Weight (g/mol) | 143.19 |
| MDL Number | MFCD00001487 |
| InChI Key | WOXWUZCRWJWTRT-UHFFFAOYSA-N |
| Synonym | 1-aminocyclohexanecarboxylic acid, homocycloleucine, 1-amino-1-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 1-amino, alpha-aminocyclohexanecarboxylic acid, alpha-aminocyclohexanecarboxylic aicd, unii-1se3hd391h, 1-amino-1-cyclohexane carboxylic acid, .alpha.-aminocyclohexanecarboxylic acid, .alpha.-aminocyclohexanecarboxylic aicd |
| PubChem CID | 1366 |
| ChEBI | CHEBI:86534 |
| IUPAC Name | 1-aminocyclohexane-1-carboxylic acid |
| SMILES | NC1(CCCCC1)C(O)=O |
Chemical Identifiers
| 2756-85-6 | |
| 143.19 | |
| WOXWUZCRWJWTRT-UHFFFAOYSA-N | |
| 1366 | |
| 1-aminocyclohexane-1-carboxylic acid |
| C7H13NO2 | |
| MFCD00001487 | |
| 1-aminocyclohexanecarboxylic acid, homocycloleucine, 1-amino-1-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 1-amino, alpha-aminocyclohexanecarboxylic acid, alpha-aminocyclohexanecarboxylic aicd, unii-1se3hd391h, 1-amino-1-cyclohexane carboxylic acid, .alpha.-aminocyclohexanecarboxylic acid, .alpha.-aminocyclohexanecarboxylic aicd | |
| CHEBI:86534 | |
| NC1(CCCCC1)C(O)=O |
Specifications
| 2756-85-6 | |
| C7H13NO2 | |
| 25 g | |
| WOXWUZCRWJWTRT-UHFFFAOYSA-N | |
| 1-aminocyclohexane-1-carboxylic acid | |
| 1366 | |
| 143.19 | |
| Crystalline Powder |
| White | |
| MFCD00001487 | |
| 1-aminocyclohexanecarboxylic acid, homocycloleucine, 1-amino-1-cyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 1-amino, alpha-aminocyclohexanecarboxylic acid, alpha-aminocyclohexanecarboxylic aicd, unii-1se3hd391h, 1-amino-1-cyclohexane carboxylic acid, .alpha.-aminocyclohexanecarboxylic acid, .alpha.-aminocyclohexanecarboxylic aicd | |
| NC1(CCCCC1)C(O)=O | |
| 143.19 | |
| CHEBI:86534 | |
| ≥98.0% (GC,T) | |
| 1-Aminocyclohexanecarboxylic Acid |
Safety and Handling
RTECSNumber : GU8393000
TSCA : Yes