missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Aminocyclohexanecarboxylic Acid 98.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 25 g |
| Formula Weight | 143.19 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1-Aminocyclohexanecarboxylic Acid |
Safety and Handling
| RTECSNumber | GU8393000 |
| TSCA | Yes |